Merck
  • Multicomponent and Multiphase Lipid Nanotubes Formed by Gliding Microtubule-Kinesin Motility and Phase-Separated Giant Unilamellar Vesicles.
Multicomponent and Multiphase Lipid Nanotubes Formed by Gliding Microtubule-Kinesin Motility and Phase-Separated Giant Unilamellar Vesicles.
Langmuir : the ACS journal of surfaces and colloids (2019-11-16)
Zachary I Imam, George D Bachand
ABSTRACT
Cytoskeletal filaments and motor proteins are critical components in the transport and reorganization of membrane-based organelles in eukaryotic cells. Previous studies have recapitulated the microtubule-kinesin transport system in vitro to dynamically assemble large-scale nanotube networks from multilamellar liposomes and polymersomes. Moving toward more biologically relevant systems, the present work examines whether lipid nanotube (LNT) networks can be generated from giant unilamellar vesicles (GUVs) and subsequently characterizes how the lipid composition may be tuned to alter the dynamics, structure, and fluidity of networks. Here, we describe a two-step process in which microtubule motility (i) drives the transport and aggregation of GUVs to form structures with a decreased energy barrier for LNT formation and (ii) extrudes LNTs without destroying parent GUVs, allowing for the formation of large LNT networks. We further show that the lipid composition of the GUV influences formation and morphology of the extruded LNTs and associated networks. For example, LNTs formed from phase-separated GUVs (e.g., liquid-solid phase-separated and coexisting liquid-ordered and liquid-disordered phase-separated) display morphologies related to the specific phase behavior reflective of the parent GUVs. Overall, the ability to form nanotubes from compositionally complex vesicles opens the door to generating lipid networks that more closely mimic the structure and function of those found in cellular systems.
Product Description
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282P, powder
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282C
Avanti
16:0 Biotinyl PE, Avanti Research - A Croda Brand 870285P, powder
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand