Merck
  • Structure before function: myosin binding protein-C slow is a structural protein with regulatory properties.
Structure before function: myosin binding protein-C slow is a structural protein with regulatory properties.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-06-07)
Janelle Geist, Christopher W Ward, Aikaterini Kontrogianni-Konstantopoulos
ABSTRACT
Myosin binding protein-C slow (sMyBP-C) comprises a family of accessory proteins in skeletal muscles that bind both myosin and actin filaments. Herein, we examined the role of sMyBP-C in adult skeletal muscles using in vivo gene transfer and clustered regularly interspaced short palindromic repeats technology to knock down all known sMyBP-C variants. Our findings, confirmed in two different skeletal muscles, demonstrated efficient knockdown (KD) of sMyBP-C (>70%) resulting in notably decreased levels of thick, but not thin, filament proteins ranging from ∼50% for slow and fast myosin to ∼20% for myomesin. Consistent with this, A bands were selectively distorted, and sarcomere length was significantly reduced. Contrary to earlier in vitro studies showing that addition of recombinant sMyBP-C slows down the formation of actomyosin crossbridges, our work demonstrates that KD of sMyBP-C in intact myofibers results in decreased contraction and relaxation kinetics under no-load conditions. Similarly, KD muscles develop markedly reduced twitch and tetanic force and contraction velocity. Taken together, our results show that sMyBP-C is essential for the regular organization and maintenance of myosin filaments into A bands and that its structural role precedes its ability to regulate actomyosin crossbridges.-Geist, J., Ward, C. W., Kontrogianni-Konstantopoulos, A. Structure before function: myosin binding protein-C slow is a structural protein with regulatory properties.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Myosin (Skeletal, Fast) antibody, Mouse monoclonal, clone MY-32, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MYBPC1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
D.E.R. 332, used as embedding medium