Skip to Content
Merck
All Photos(1)

Key Documents

31185

Sigma-Aldrich

D.E.R. 332

used as embedding medium

Synonym(s):

Bisphenol A diglycidyl ether, Epoxy-Resin, 2,2-Bis[4-(glycidyloxy)phenyl]propane, 4,4′-Isopropylidenediphenol diglycidyl ether, BADGE, D.E.R. 332

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O4
CAS Number:
Molecular Weight:
340.41
Beilstein:
299026
EC Number:
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.25

SMILES string

CC(C)(c1ccc(OCC2CO2)cc1)c3ccc(OCC4CO4)cc3

InChI

1S/C21H24O4/c1-21(2,15-3-7-17(8-4-15)22-11-19-13-24-19)16-5-9-18(10-6-16)23-12-20-14-25-20/h3-10,19-20H,11-14H2,1-2H3

InChI key

LCFVJGUPQDGYKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The uniqueness of D.E.R. 332 epoxy resin is reflected in its maximum epoxide equivalent weight of 178. Because of its high purity and lack of polymer factions, samples embedded using D.E.R. 322 have uniform performance, exceptionally low viscosity and color; and improved elevated temperature properties.

Caution

Crystallizes very slowly at room-temperature; can be melted by heating up to 50-55 °C

Legal Information

D.E.R. is a trademark of The Dow Chemical Company or an affiliated company of Dow

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

507.2 - 514.4 °F - closed cup

Flash Point(C)

264 - 268 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Light and electron microscopy of diet-induced atrial thrombosis in TS mice. I. Primary changes in the endocardium.
W R Lockwood et al.
The American journal of anatomy, 126(2), 185-199 (1969-10-01)
Zihan Xu et al.
Developmental and comparative immunology, 100, 103413-103413 (2019-06-15)
The objective of this study is to explore the immune response of the shrimp Penaeus vannamei to low temperature and air exposure during the mimic waterless transportation. Shrimp were cold shocked at 13 °C for 3 min, then exposed to air at
Wen-Jie Cheng et al.
Life sciences, 231, 116533-116533 (2019-06-08)
The aim of this study was to investigate the protective effect of chronic intermittent hypobaric hypoxia (CIHH) against skeletal muscle ischemia-reperfusion (IR) injury and to determine the underlying mechanism. C57BL/6 mice were randomly divided into 3 groups: skeletal muscle IR
Miroslav Müller et al.
Polymers, 11(7) (2019-07-03)
This paper deals with a research focused on utilization of microparticle and short-fiber filler based on cotton post-harvest line residues in an area of polymeric composites. Two different fractions of the biological filler (FCR-reinforced cotton filler) of 20 and 100
Ayşe Aydoğmuş Erik et al.
Microscopy research and technique, 82(9), 1448-1454 (2019-05-24)
This experimental study aims to evaluate the radiopacity of various fiber post materials and to determine the effects of material composition as analyzed by energy-dispersive X-ray spectrophotometry (EDS; EDAX Team Software; EDAX, Inc., Mahwah, NJ) on radiopacity. Five specimens of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service