Merck
  • Reorganization of Ternary Lipid Mixtures of Nonphosphorylated Phosphatidylinositol Interacting with Angiomotin.
Reorganization of Ternary Lipid Mixtures of Nonphosphorylated Phosphatidylinositol Interacting with Angiomotin.
The journal of physical chemistry. B (2018-06-08)
Ann C Kimble-Hill, Horia I Petrache, Soenke Seifert, Millicent A Firestone
ABSTRACT
Phosphatidylinositol (PI) lipids are necessary for many cellular signaling pathways of membrane associated proteins, such as angiomotin (Amot). The Amot family regulates cellular polarity, growth, and migration. Given the low concentration of PI lipids in these membranes, it is likely that such protein-membrane interactions are stabilized by lipid domains or small lipid clusters. By small-angle X-ray scattering, we show that nonphosphorylated PI lipids induce lipid demixing in ternary mixtures of phosphatidylcholine (PC) and phosphatidylethanolamine (PE), likely because of preferential interactions between the head groups of PE and PI. These results were obtained in the presence of buffer containing tris(hydroxymethyl)aminomethane, 4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid, NaCl, ethylenediaminetetraacetic acid, dithiothreitol, and benzamidine at pH 8.0 that in previous work showed an ability to cause PC to phase separate but are necessary to stabilize Amot for in vitro experimentation. Collectively, this provided a framework for determining the effect of Amot on lipid organization. Using fluorescence spectroscopy, we were able to show that the association of Amot with this lipid platform causes significant reorganization of the lipid into a more homogenous structure. This reorganization mechanism could be the basis for Amot membrane association and fusogenic activity previously described in the literature and should be taken into consideration in future protein-membrane interaction studies.
Product Description
Avanti
TopFluor TMR PI, Avanti Research - A Croda Brand 810188P, powder
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
Soy PI, Avanti Research - A Croda Brand
Avanti
Soy PI, Avanti Research - A Croda Brand 840044C
Avanti
TopFluor TMR PE, Avanti Research - A Croda Brand 810241C
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131C
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131P, powder
Avanti
TopFluor TMR PE, Avanti Research - A Croda Brand 810241P, powder
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand