Merck
  • A Flat BAR Protein Promotes Actin Polymerization at the Base of Clathrin-Coated Pits.
A Flat BAR Protein Promotes Actin Polymerization at the Base of Clathrin-Coated Pits.
Cell (2018-06-12)
Leonardo Almeida-Souza, Rene A W Frank, Javier García-Nafría, Adeline Colussi, Nushan Gunawardana, Christopher M Johnson, Minmin Yu, Gillian Howard, Byron Andrews, Yvonne Vallis, Harvey T McMahon
ABSTRACT
Multiple proteins act co-operatively in mammalian clathrin-mediated endocytosis (CME) to generate endocytic vesicles from the plasma membrane. The principles controlling the activation and organization of the actin cytoskeleton during mammalian CME are, however, not fully understood. Here, we show that the protein FCHSD2 is a major activator of actin polymerization during CME. FCHSD2 deletion leads to decreased ligand uptake caused by slowed pit maturation. FCHSD2 is recruited to endocytic pits by the scaffold protein intersectin via an unusual SH3-SH3 interaction. Here, its flat F-BAR domain binds to the planar region of the plasma membrane surrounding the developing pit forming an annulus. When bound to the membrane, FCHSD2 activates actin polymerization by a mechanism that combines oligomerization and recruitment of N-WASP to PI(4,5)P2, thus promoting pit maturation. Our data therefore describe a molecular mechanism for linking spatiotemporally the plasma membrane to a force-generating actin platform guiding endocytic vesicle maturation.
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
3-sn-Phosphatidylethanolamine from bovine brain, Type I, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Anti-ARP3 antibody, Mouse monoclonal, clone FMS338, purified from hybridoma cell culture
Avanti
18:0-20:4 PI(3,4,5)P3, 1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human FCHSD2
Sigma-Aldrich
MISSION® esiRNA, targeting human ITSN1
Sigma-Aldrich
MISSION® esiRNA, targeting human ITSN2
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
1,2-Diacyl-sn-glycero-3-phospho-L-serine solution, ≥97%, from bovine brain, chloroform:methanol solution
Sigma-Aldrich
Brain Extract from bovine brain, Type I, Folch Fraction I
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Avanti
08:0 PI(3,4)P2, 1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′-bisphosphate) (ammonium salt), powder