Merck
  • NUPR1 maintains autolysosomal efflux by activating SNAP25 transcription in cancer cells.
NUPR1 maintains autolysosomal efflux by activating SNAP25 transcription in cancer cells.
Autophagy (2017-11-14)
Yanchao Mu, Xiaojie Yan, Ding Li, Dan Zhao, Lingling Wang, Xiaoyang Wang, Dan Gao, Jie Yang, Hua Zhang, Yanzhe Li, Yanan Sun, Yiliang Wei, Zhenfa Zhang, Xinzhong Chang, Zhi Yao, Shanshan Tian, Kai Zhang, Lance S Terada, Zhenyi Ma, Zhe Liu
ABSTRACT
In the advanced stages of cancer, autophagy is thought to promote tumor progression through its ability to mitigate various cellular stresses. However, the details of how autophagy is homeostatically regulated in such tumors are unknown. Here, we report that NUPR1 (nuclear protein 1, transcriptional regulator), a transcriptional coregulator, is aberrantly expressed in a subset of cancer cells and predicts low overall survival rates for lung cancer patients. NUPR1 regulates the late stages of autolysosome processing through the induction of the SNARE protein SNAP25, which forms a complex with the lysosomal SNARE-associated protein VAMP8. NUPR1 depletion deregulates autophagic flux and impairs autolysosomal clearance, inducing massive cytoplasmic vacuolization and premature senescence in vitro and tumor suppression in vivo. Collectively, our data show that NUPR1 is a potent regulator of autolysosomal dynamics and is required for the progression of some epithelial cancers.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Goat serum
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, laboratory grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
SKU
Pack Size
Availability
Price
Quantity
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
SKU
Pack Size
Availability
Price
Quantity
Millipore
ProteinA-Sepharose 4, Fast Flow from Staphylococcus aureus, aqueous ethanol suspension
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human NUPR1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Normal Rat IgG, This Normal Rat IgG is validated for use in ELISA Flow Cytometry Immunoblotting Immunofluorescence Immunohistochemistry Immunoprecipitation for the detection of Rat IgG, Normal.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O