Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tributylamine
Synonym(s):
Tributylamine
Linear Formula:
[CH
3
(CH
2
)
3
]
3
N
CAS Number:
102-82-9
Molecular Weight:
185.35
EC Number:
203-058-7
Products
Product Category
solvents (1)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (2)
Puriss P.A. (1)
Puriss Plus (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
471313
Tributylamine,
≥98.5%
Expand
View Pricing
90781
Tributylamine,
puriss. plus, ≥99.5% (GC)
Expand
View Pricing
8.08358
Tributylamine,
for synthesis
Expand
View Pricing
90780
Tributylamine,
puriss. p.a., ≥99.0% (GC)
Expand
View Pricing