Skip to Content
Merck
All Photos(1)

Key Documents

8.08358

Sigma-Aldrich

Tributylamine

for synthesis

Synonym(s):

Tributylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)3N
CAS Number:
Molecular Weight:
185.35
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-058-7
NACRES:
NA.22
Assay:
≥99.0% (GC)
bp:
216-217 °C/1013 hPa
vapor pressure:
0.18 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

0.18 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

190 °C

potency

420-780 mg/kg LD50, oral (Rat)
195 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-6.0 % (v/v)

pH

10.6 (20 °C in H2O, saturated aqueous solution)

bp

216-217 °C/1013 hPa

mp

-70 °C

transition temp

flash point 63-66 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCCC)(CCCC)CCCC

InChI

1S/C12H27N/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

IMFACGCPASFAPR-UHFFFAOYSA-N

Application


  • Temperature Effects on the Thermodynamic Parameters of Tributylamine Adsorption on Carbon Steel as a Corrosion Inhibitor: This study explores tributylamine′s effectiveness as a corrosion inhibitor for carbon steel in carbon dioxide-saturated environments, optimizing inhibitor concentration for maximum efficiency (Hashemi et al., 2023).

  • Method development for the acquisition of adsorption isotherm of ion pair reagents Tributylamine and Triethylamine in ion pair chromatography: Investigates the optimization of ion pair chromatography for analyzing oligonucleotides using tributylamine and triethylamine, emphasizing methodological improvements in chromatographic analysis (Haseeb et al., 2023).


Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.776 - 0.777
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service