![Ropivacaine hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/083/68f5c9d2-46df-4bc3-b6c1-77e19aebc0b5/640/68f5c9d2-46df-4bc3-b6c1-77e19aebc0b5.png)
Ropivacaine hydrochloride
![Ropivacaine hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/083/68f5c9d2-46df-4bc3-b6c1-77e19aebc0b5/640/68f5c9d2-46df-4bc3-b6c1-77e19aebc0b5.png)
Synonym(s):
Ropivacaine hydrochloride monohydrate, (2S)-N-(2,6-dimethylphenyl)-1-propyl-2-piperidinecarboxamide hydrochloride monohydrate, (S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrate, Naropin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C17H26N2O · HCl · H2O
CAS Number:
Molecular Weight:
328.88
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing