
N,N′,N′′-Triacetylchitotriose

Synonym(s):
O-[2-Acetamido-2-deoxy-β-D-glucopyranosyl-(1→4)]-O-[2-acetamido-2-deoxy-β-D-glucopyranosyl-(1→4)]-2-acetamido-2-deoxy-D-glucopyranose
Empirical Formula (Hill Notation):
C24H41N3O16
CAS Number:
Molecular Weight:
627.59
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1