
Glyceryl triundecanoate

Synonym(s):
1,2,3-Triundecanoylglycerol, Glycerol triundecanoate, Triundecanoin
Linear Formula:
[CH3(CH2)9COOCH2]2CHOCO(CH2)9CH3
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1