
Fenoterol hydrobromide

Synonym(s):
2-(3,5-Dihydroxyphenyl)-2-hydroxy-2′-(4-hydroxyphenyl)-1′-methyldiethylamine hydrobromide
Empirical Formula (Hill Notation):
C17H21NO4 · HBr
CAS Number:
Molecular Weight:
384.26
Products
Product Category
Formula Weight
Gene Alias
Manufacturer
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing