Skip to Content
Merck
Azithromycin Related Compound H certified reference material, pharmaceutical secondary standard

Azithromycin Related Compound H

Azithromycin Related Compound H certified reference material, pharmaceutical secondary standard
Synonym(s):
3′-N-[[4-(Acetylamino)phenyl]sulfonyl]-3′-N-demethylazithromycin
Empirical Formula (Hill Notation):
C45H77N3O15S
CAS Number:
Molecular Weight:
932.17

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2