
Acifluorfen

Synonym(s):
5-(2-Chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoic acid, 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid
Empirical Formula (Hill Notation):
C14H7ClF3NO5
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2