Skip to Content
Merck
6-Acetylmorphine, 1.0 mg/mL solution 1 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

6-Acetylmorphine, 1.0 mg/mL solution

6-Acetylmorphine, 1.0 mg/mL solution 1 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
6-Acetylmorphine solution
Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique