Skip to Content
Merck
All Photos(1)

Key Documents

1724500

USP

Zidovudine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3′-Azido-3′-deoxythymidine, AZT, Azidothymidine, ZDV, Zidovudine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
Beilstein:
3595791
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

zidovudine

manufacturer/tradename

USP

mp

113-115 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H13N5O4/c1-5-3-15(10(18)12-9(5)17)8-2-6(13-14-11)7(4-16)19-8/h3,6-8,16H,2,4H2,1H3,(H,12,17,18)/t6-,7+,8+/m0/s1

InChI key

HBOMLICNUCNMMY-XLPZGREQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Zidovudine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Abacavir, Lamivudine and Zidovudine Tablets
  • Lamivudine and Zidovudine Tablets
  • Zidovudine
  • Zidovudine Capsules
  • Zidovudine Injection
  • Zidovudine Oral Solution
  • Zidovudine Tablets

Biochem/physiol Actions

Reverse transcriptase inhibitor active against HIV-1 virus.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y-H Lai et al.
Gene therapy, 22(2), 155-162 (2014-10-31)
Replicating virus vectors are attractive tools for anticancer gene therapy, but the potential for adverse events due to uncontrolled spread of the vectors has been a major concern. To design a tumor-specific retroviral replicating vector (RRV), we replaced the U3
Youko Suehiro et al.
British journal of haematology, 169(3), 356-367 (2015-01-24)
Adult T cell leukaemia/lymphoma (ATL) is a human T cell leukaemia virus type-I (HTLV-I)-infected T cell malignancy with poor prognosis. We herein developed a novel therapeutic vaccine designed to augment an HTLV-I Tax-specific cytotoxic T lymphocyte (CTL) response that has
H G Sprenger et al.
HIV medicine, 16(2), 122-131 (2014-12-05)
The aim of the study was to test the antiviral efficacy of a triple nucleoside reverse transcriptase inhibitor (NRTI) regimen, with potential beneficial metabolic effects, as maintenance therapy after induction with dual NRTIs and a boosted protease inhibitor (PI). An
Richard M Hoglund et al.
British journal of clinical pharmacology, 79(4), 636-649 (2014-10-10)
Drug-drug interactions between antimalarial and antiretroviral drugs may influence antimalarial treatment outcomes. The aim of this study was to investigate the potential drug-drug interactions between the antimalarial drugs, lumefantrine, artemether and their respective metabolites desbutyl-lumefantrine and dihydroartemisinin, and the HIV
H Stedt et al.
Cancer gene therapy, 22(3), 130-137 (2015-01-24)
Malignant gliomas (MGs) are the most common malignant primary brain tumors with a short life estimate accompanied by a marked reduction in the quality of life. Herpes Simplex virus-1 thymidine kinase ganciclovir (HSV-TK/GCV) system is the best characterized enzyme prodrug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service