Skip to Content
Merck
All Photos(1)

Key Documents

1709007

USP

Vancomycin hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Vancomycin monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · HCl
Molecular Weight:
1485.71
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vancomycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

Vancomycin Hydrochloride is the hydrochloride salt of a kind of vancomycin, a substance produced by the growth of Streptomyces orientalis (Fam. Streptomycetaceae), or a mixture of two or more such salts.
Vancomycin Hydrochloride USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vancomycin hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Vancomycin Hydrochloride Capsules
  • Vancomycin Hydrochloride for Injection
  • Vancomycin Hydrochloride for Oral Solution
  • Vancomycin Injection
  • Vancomycin

Packaging

4 vials, each vial contains 99,300 mcg of vancomycin activity

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vancomycin Hydrochloride for Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 35(4), 4588-4588 (2020)
Vancomycin Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 27(4), 4585-4585 (2022)
Vancomycin
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(5), 4583-4583 (2020)
Vancomycin Hydrochloride for Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(1), 4590-4590 (2013)
Vancomycin Hydrochloride Capsules
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 34(6), 4588-4588 (2020)

Related Content

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service