Skip to Content
Merck

1553815

USP

Pramipexole Related Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dexpramipexole dihydrochloride, (R)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole dihydrochloride // (R)-Pramipexole dihydrochloride, (R)-4,5,6,7-Tetrahydro-6-(propylamino)-benzothiazole-2-amine dihydrochloride, (R)-Pramipexole dihydrochloride, KNS 760704, R-(+)-Pramipexole dihydrochloride, RPPX, SND 919CL2X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H17N3S · 2HCl
CAS Number:
Molecular Weight:
284.25
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

pramipexole

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.Cl.CCCN[C@@H]1CCc2nc(N)sc2C1

InChI

1S/C10H17N3S.2ClH/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;/h7,12H,2-6H2,1H3,(H2,11,13);2*1H/t7-;;/m1../s1

InChI key

QMNWXHSYPXQFSK-XCUBXKJBSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pramipexole Related Compound D USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

R-(+)-Pramipexole is a neuroprotective agent; weak non-ergoline dopamine agonist.
R-(+)-Pramipexole is a neuroprotective agent; weak non-ergoline dopamine agonist. R-(+)-Pramipexole has been found to have neuroprotective effects and is being investigated for treatment of ALS. It reduces mitochondrial reactive oxygen species (ROS) production, inhibits the activation of apoptotic pathways, and increase cell survival in response to a variety of neurotoxins and β-amyloid neurotoxicity. Compared to the S-(-) isomer, R-(+)-Pramipexole has much lower dopamine agonist activity.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Pramipexole Related Compound D


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library