Skip to Content
Merck

1466607

USP

N-Nitrosomethylphenylamine (NMPA)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Methyl-N-nitrosoaniline, N-Methyl-N-nitrosobenzenamine, N-Methyl-N-phenylnitrosamine, NMPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

packaging

pkg of 1 mL

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

N(N=O)(C)c1ccccc1

InChI

1S/C7H8N2O/c1-9(8-10)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

MAXCWSIJKVASQC-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Nitrosamines are chemical compounds that can form during drug manufacturing and may pose health risks, including carcinogenicity.

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.

Application

N-Nitrosomethylphenylamine (NMPA) USP reference standard is intended for use only as specifically prescribed in the United States Pharmacopoeia.

It is also used to prepare standard, and sensitivity solutions to determine NMPA impurity in drug substances and drug products (valsartan, irbesartan, and losartan potassium etc) by chromatography according to general chapter 〈1469〉 of United States Pharmacopeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under N-Nitrosomethylphenylamine (NMPA)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

49.5 °F

flash_point_c

9.7 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library