Skip to Content
Merck
All Photos(1)

Key Documents

1463701

USP

Nifedipine Nitrosophenylpyridine Analog

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O5
CAS Number:
Molecular Weight:
328.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nifedipine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C(OC)C1=C(C2=C(N=O)C=CC=C2)C(C(OC)=O)=C(C)N=C1C

InChI

1S/C17H16N2O5/c1-9-13(16(20)23-3)15(11-7-5-6-8-12(11)19-22)14(10(2)18-9)17(21)24-4/h5-8H,1-4H3

InChI key

MUZLTKGYFZENFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nifedipine Nitrosophenylpyridine Analog USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Nifedipine
  • Nifedipine Capsules
  • Nifedipine Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Carlucci et al.
Farmaco (Societa chimica italiana : 1989), 45(6 Suppl), 751-755 (1990-06-01)
A method for the determination of 2,6-dimethyl-4-(2'-nitrosophenyl)-3,5-pyridinedicarboxylic acid dimethylester in nifedipine (bulk material and pharmaceutical formulations) by second-derivative ultraviolet spectrophotometry is described. The procedure is simple and rapid and gives accurate and precise results.
Jürgen Schoppe et al.
Cell calcium, 33(3), 207-221 (2003-02-26)
Leech P neurons possess caffeine-sensitive ion channels in intracellular Ca(2+) stores and in the plasma membrane. The following results indicate that these channels are also activated by 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester (NTP), the photoproduct of the L-type Ca(2+) channel-blocker nifedipine:
V Misík et al.
Molecular pharmacology, 40(3), 435-439 (1991-09-01)
Nifedipine [1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic+ ++ acid dimethyl ester] and nimodipine [1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic+ ++ acid 2-methoxyethyl 1-methylethyl ester], incorporated into diheptanoylphosphatidylcholine liposomes, which were used as a drug carrier system, slightly inhibited lipid peroxidation (induced by tert-butylhydroperoxide and Fe2+) in rat heart homogenate. Illumination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service