Skip to Content
Merck

Skip To

1454008

USP

Nandrolone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

19-Nortestosterone, 17β-Hydroxy-19-norandrost-4-en-3-one, 17β-Hydroxy-4-estren-3-one, 19-Norandrostenolone, 4-Estren-17β-ol-3-one, Nadrolone, Nandrolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H26O2
CAS Number:
Molecular Weight:
274.40
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2055849
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nandrolone

manufacturer/tradename

USP

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1

InChI key

NPAGDVCDWIYMMC-IZPLOLCNSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nandrolone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Nandrolone Decanoate
  • Nandrolone Decanoate Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP2571455000Y0000542
Nandrolone British Pharmacopoeia (BP) Reference Standard

BP257

Nandrolone

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

USP

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

drug control

-

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

drug control

-

API family

nandrolone

API family

nandrolone

API family

nandrolone

API family

nandrolone


Still not finding the right product?

Explore all of our products under Nandrolone


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters