Skip to Content
Merck
All Photos(1)

Key Documents

1430349

USP

Methyl linolenate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Linolenic acid methyl ester, Methyl cis,cis,cis-9,12,15-octadecatrienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)7COOCH3
CAS Number:
Molecular Weight:
292.46
Beilstein:
1728194
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methyl linolenate

manufacturer/tradename

USP

refractive index

n20/D 1.470 (lit.)

bp

182 °C/3 mmHg (lit.)

density

0.895 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h4-5,7-8,10-11H,3,6,9,12-18H2,1-2H3/b5-4-,8-7-,11-10-

InChI key

DVWSXZIHSUZZKJ-YSTUJMKBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methyl linolenate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Borage Seed Oil Capsules
  • Evening Primrose Oil Capsules
  • Flax Seed Oil Capsules
  • Powdered Saw Palmetto
  • Saw Palmetto
  • Saw Palmetto Capsules
  • Saw Palmetto Extract

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service