Skip to Content
Merck
All Photos(1)

Key Documents

1380105

USP

Melatonin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Acetyl-5-methoxytryptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
Beilstein:
205542
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

melatonin

manufacturer/tradename

USP

mp

116.5-118 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]cc(CCNC(C)=O)c2c1

InChI

1S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15H,5-6H2,1-2H3,(H,14,16)

InChI key

DRLFMBDRBRZALE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Melatonin USP reference standard for use in specified quality tests and assays.

Also used to prepare standard and system suitability solutions for assay, performance tests, strength, and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Melatonin Tablets
  • Melatonin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melatonin
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(4), 5143-5143 (2020)
Melatonin Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(6), 5144-5144 (2013)
Ismail Gögenur et al.
Journal of pineal research, 57(1), 10-15 (2014-04-09)
The aim was to examine the effect of perioperative melatonin treatment on clinical cardiac morbidity and markers of myocardial ischemia in patients undergoing elective surgery for abdominal aortic aneurism. Reperfusion injury results in increased cardiac morbidity in patients undergoing surgery
Reuben K Wong et al.
Digestive diseases and sciences, 60(1), 186-194 (2014-08-06)
Probiotics have treatment efficacy in irritable bowel syndrome (IBS), but the exact mechanism remains obscure. One hypothesis is the mediation of melatonin levels, leading to changes in IBS symptoms. The purpose of this study was to evaluate the effects of
Mariano Bizzarri et al.
Expert opinion on therapeutic targets, 17(12), 1483-1496 (2013-09-17)
Compelling evidence has highlighted the complex pleiotropic functions elicited by the melatonin in cancer cells. Melatonin behaves as a 'smart killer', i.e., modulating anti-apoptotic processes in normal cells, and triggering pro-apoptotic signals in cancer cells. Melatonin induces programmed cell death

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service