Skip to Content
Merck

1347880

USP

Isobutyraldehyde

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methylpropanal, 2-Methylpropionaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
605330
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

2.5 (vs air)

vapor pressure

66 mmHg ( 4.4 °C)

autoignition temp.

384 °F

expl. lim.

10 %, 25 °F, 2 %, 32 °F

manufacturer/tradename

USP

refractive index

n20/D 1.374 (lit.)

bp

63 °C (lit.)

mp

−65 °C (lit.)

density

0.79 g/mL at 25 °C (lit.)

application(s)

pharmaceutical
pharmaceutical (small molecule)

SMILES string

[H]C(=O)C(C)C

InChI

1S/C4H8O/c1-4(2)3-5/h3-4H,1-2H3

InChI key

AMIMRNSIRUDHCM-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isobutyraldehyde USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Isobutyl Alcohol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Isobutyraldehyde


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-11.2 °F - closed cup

flash_point_c

-24 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library