Skip to Content
Merck

Skip To

1333058

USP

Hydroxyzine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-(4-Chlorobenzhydryl)piperazine, (RS)-1-[(4-Chlorophenyl)phenylmethyl]piperazine, NSC 86164, Norchlorcyclizine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H19ClN2
CAS Number:
Molecular Weight:
286.80
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

hydroxyzine

manufacturer/tradename

USP

bp

178-180 °C/0.5 mmHg (lit.)

mp

65-70 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccc(cc1)C(N2CCNCC2)c3ccccc3

InChI

1S/C17H19ClN2/c18-16-8-6-15(7-9-16)17(14-4-2-1-3-5-14)20-12-10-19-11-13-20/h1-9,17,19H,10-13H2

InChI key

UZKBSZSTDQSMDR-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxyzine Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Hydroxyzine Pamoate
  • Hydroxyzine Pamoate Capsules
  • Hydroxyzine Hydrochloride
  • Hydroxyzine Hydrochloride Oral Solution
  • Hydroxyzine Hydrochloride Tablets
  • Cetirizine Hydrochloride Orally Disintegrating Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR287211029301643463
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

-

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

hydroxyzine

API family

hydroxyzine

API family

cetirizine

API family

terazosin

bp

178-180 °C/0.5 mmHg (lit.)

bp

178-180 °C/0.5 mmHg (lit.)

bp

-

bp

-


Still not finding the right product?

Explore all of our products under Hydroxyzine Related Compound A


signalword

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters