1287653
USP
Gadoteridol Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(1,4,7,10-Tetraazacyclododecane-1,4,7-triacetato(3-)-N1,N4,N7,N10,O1,O4,O7)gadolinium, 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic acid monogadolinium salt, Des(2-hydroxypropyl) gadoteridol
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
gadoteridol
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
[Gd+3].[N-]1CCN(CCN(CCN(CC1)CC(=O)[O-])CC(=O)[O-])CC(=O)[O-]
InChI
1S/C14H25N4O6.Gd/c19-12(20)9-16-3-1-15-2-4-17(10-13(21)22)6-8-18(7-5-16)11-14(23)24;/h1-11H2,(H,19,20)(H,21,22)(H,23,24);/q-1;+3/p-3
InChI key
VXYQJCHCLZZIRW-UHFFFAOYSA-K
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service