Skip to Content
Merck
All Photos(1)

Key Documents

1287303

USP

Gabapentin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-(Aminomethyl)-cyclohexaneacetic acid, Neurontin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
Beilstein:
2359739
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

gabapentin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NCC1(CC(O)=O)CCCCC1

InChI

1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)

InChI key

UGJMXCAKCUNAIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gabapentin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Gabapentin Capsules
  • Gabapentin Tablets
  • Gabapentin Compounded Cream
  • Gabapentin Compounded Oral Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ESTIMATION OF GABAPENTIN IN HUMAN PLASMA USING LC-MS/MS METHOD.
Das, Binaya.
Asian Journal of Pharmaceutical and Clinical Research, 6.3, 213-216 (2013)
N S Gee et al.
The Journal of biological chemistry, 271(10), 5768-5776 (1996-03-08)
Gabapentin (1-(aminomethyl)cyclohexane acetic acid; Neurontin) is a novel anticonvulsant drug, with a mechanism of action apparently dissimilar to that of other antiepileptic agents. We report here the isolation and characterization of a [3H]gabapentin-binding protein from pig cerebral cortex membranes. The
Nada Alayed et al.
Obstetrics and gynecology, 123(6), 1221-1229 (2014-05-09)
To examine the evidence of preemptive use of gabapentin in abdominal hysterectomy. We conducted an electronic based search using the following databases: PubMed, EMBASE, Ovid MEDLINE, ClinicalTrials.gov, and Cochrane Central Register of Controlled Trials. The following medical subject heading terms
Cuiping Chen et al.
Journal of pharmaceutical sciences, 102(4), 1155-1164 (2013-02-06)
Neurontin®, an immediate-release (IR) formulation of gabapentin, was the first drug approved by the United States Food and Drug Administration for the treatment of postherpetic neuralgia (PHN). The effective dosing regimen of gabapentin IR (G-IR) for PHN is 1800 mg/day in
Kim Abbass
The Annals of pharmacotherapy, 46(12), 1707-1711 (2012-12-13)
To evaluate the effectiveness of gabapentin in adults with phantom limb pain. A PubMed search (1966-September 2012) was conducted using the key words phantom limb pain and gabapentin. Search limits were English language, humans, adult, clinical trials, and randomized controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service