Skip to Content
Merck
All Photos(1)

Key Documents

1287030

USP

Furosemide Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Chloro-5-sulfamoylanthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClN2O4S
CAS Number:
Molecular Weight:
250.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

furosemide

manufacturer/tradename

USP

mp

267 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1cc(Cl)c(cc1C(O)=O)S(N)(=O)=O

InChI

1S/C7H7ClN2O4S/c8-4-2-5(9)3(7(11)12)1-6(4)15(10,13)14/h1-2H,9H2,(H,11,12)(H2,10,13,14)

InChI key

QQLJBZFXGDHSRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Furosemide Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Nakahama et al.
European journal of clinical pharmacology, 32(3), 313-315 (1987-01-01)
Urine from 5 renal transplant recipients treated with frusemide was analyzed for unchanged frusemide (F), glucuronidated frusemide (G) and 4-chloro-5-sulfamoylanthranilic acid (CSA) by HPLC. In 3 recipients, whose renal function recovered steadily and whose hepatic function was normal throughout, the
M Hezari et al.
Drug metabolism and disposition: the biological fate of chemicals, 20(6), 882-888 (1992-11-01)
Furosemide (Lasix), a widely used diuretic, is metabolized by the fungus Cunninghamella elegans (ATCC 36112) to 4-chloro-5-sulfamoyl anthranilic acid (CSA), a metabolite also present in mammalian systems. This metabolite was isolated following preparative-scale incubations of C. elegans, and was characterized
D E Smith et al.
Drug metabolism and disposition: the biological fate of chemicals, 8(5), 337-342 (1980-09-01)
The objectives of this study were to qualitatively and quantitatively compare the metabolism, pharmacokinetics, and bioavailability of furosemide in healthy volunteers after intravenous and oral administration. We also determined the plasma protein binding of furosemide in vivo after iv administration.
[Determination of furosemide and its metabolic products in plasma and urine by high performance liquid chromatography and clinical application].
Y Miwa et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 108(11), 1087-1092 (1988-11-01)
L Z Benet et al.
Federation proceedings, 42(6), 1695-1698 (1983-04-01)
A variety of furosemide assays and their application to biological samples have been described over the past 2 decades. We describe development of a specific, sensitive assay appropriate for efficient measurement of large numbers of biological samples. With development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service