Skip to Content
Merck

1269152

USP

Famciclovir

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(2-(2-Amino-9H-purin-9-yl)ethyl)-1,3-propanediol diacetate ester, BRL 42810

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H19N5O4
CAS Number:
Molecular Weight:
321.33
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

famciclovir

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OCC(CCn1cnc2cnc(N)nc12)COC(C)=O

InChI

1S/C14H19N5O4/c1-9(20)22-6-11(7-23-10(2)21)3-4-19-8-17-12-5-16-14(15)18-13(12)19/h5,8,11H,3-4,6-7H2,1-2H3,(H2,15,16,18)

InChI key

GGXKWVWZWMLJEH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Famciclovir USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Famciclovir Tablets
  • Famciclovir Compounded Oral Suspension

Biochem/physiol Actions

Famciclovir is an Antiviral; Guanine analog used to treat herpesvirus infections.
Famciclovir is an antiretroviral guanosine analog used to treat herpesvirus infections and hepatitis B. Famciclovir is rapidly converted to penciclovir. Viral thymidine kinase phosphorylates penciclovir to a monophosphate form that celular kinases convert in turn to penciclovir triphosphate. Penciclovir triphosphate competitively inhibits viral DNA polymerase and thus viral replication. Prolonged administration can lead to resistance; it is often manifested as selection of pre-existing resistant strains with mutations in the reverse transcriptase domain of the DNA polymerase gene.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Famciclovir


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library