Skip to Content
Merck
All Photos(1)

Key Documents

1236970

USP

Epinephrine

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

epinephrine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Epinephrine is classified as sympathomimetic catecholamine. It is a stress hormone and functions as part of the ′fight or flight′ response.

Application

Epinephrine USP reference standard intended for use in specified quality tests and assays.

Also used to prepare standard, sensitivity, and system suitability solutions during the assay and impurity analysis of Epinephrine by using liquid chromatography in conjunction with a UV detector.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epinephrine
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 1645-1645 (2020)
Endocrinology of stress
Romero, Michael L and Butler, Luke K
Journal of Comparative Psychology (Washington, D.C. : 1983), 20(2) (2007)
Sequential determination of hydrocortisone and epinephrine in pharmaceutical formulations via photochemically enhanced fluorescence
Cardoso CE, et al.
Microchimica Acta, 146(1), 79-84 (2004)
Automated flow-injection spectrophotometric determination of catecholamines (epinephrine and isoproterenol) in pharmaceutical formulations based on ferrous complex formation.
Solich P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 22(5), 781-789 (2000)
Separation and determination of norepinephrine, epinephrine and isoprinaline enantiomers by capillary electrophoresis in pharmaceutical formulation and human serum
Wei S, et al.
Journal of Chromatography A, 1098(1-2), 166-171 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service