Skip to Content
Merck
All Photos(1)

Documents

1235300

USP

Enalapril maleate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Enalapril maleate salt, (S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N2O5 · C4H4O4
CAS Number:
Molecular Weight:
492.52
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

enalapril

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@@H](C(N1[C@H](C(O)=O)CCC1)=O)N[C@H](CC(OCC)=O)CCC2=CC=CC=C2.O=C(OO)/C=C\C(O)=O

InChI

1S/C21H30N2O5.C4H4O5/c1-3-28-19(24)14-17(12-11-16-8-5-4-6-9-16)22-15(2)20(25)23-13-7-10-18(23)21(26)27;5-3(6)1-2-4(7)9-8/h4-6,8-9,15,17-18,22H,3,7,10-14H2,1-2H3,(H,26,27);1-2,8H,(H,5,6)/b;2-1-/t15-,17-,18-;/m0./s1

InChI key

UQKQUZOIQFOJOI-KJHLASMFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enalapril maleate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Enalapril Maleate Tablets
  • Enalapril Maleate and Hydrochlorothiazide Tablets
  • Enalapril Maleate Compounded Oral Suspension
  • Enalaprilat Injection

Biochem/physiol Actions

A long-acting angiotensin-converting enzyme inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Heart failure and LCZ696].
Guy Sabourin
Perspective infirmiere : revue officielle de l'Ordre des infirmieres et infirmiers du Quebec, 12(1), 61-61 (2015-02-06)
John J V McMurray et al.
The New England journal of medicine, 371(11), 993-1004 (2014-09-02)
We compared the angiotensin receptor-neprilysin inhibitor LCZ696 with enalapril in patients who had heart failure with a reduced ejection fraction. In previous studies, enalapril improved survival in such patients. In this double-blind trial, we randomly assigned 8442 patients with class
Neprilysin inhibition--a novel therapy for heart failure.
Mariell Jessup
The New England journal of medicine, 371(11), 1062-1064 (2014-09-02)
PARADIGM-HF--the experts' discussion.
Mariell Jessup et al.
The New England journal of medicine, 371(11), e15-e15 (2014-09-04)
Marta de Diego et al.
Journal of chromatographic science, 52(10), 1193-1197 (2013-12-04)
A simple and rapid stability-indicating liquid chromatographic method was developed and validated for the simultaneous determination of lisinopril and hydrochlorotiazide (HCTZ) in drug substances and dosage forms in the presence of degradation products. Forced degradation studies were conducted on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service