Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
API family
cinnamaldehyde
manufacturer/tradename
USP
application(s)
pharmaceutical
SMILES string
O=C\C=C\c1ccccc1
InChI
1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+
InChI key
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Cinnamaldehyde USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Cinnamomum cassia Bark Powder
- Cinnamomum cassia Bark
- Cinnamomum cassia Twig
- Cinnamomum cassia Twig Powder
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Cinnamaldehyde
signalword
Warning
Storage Class
10 - Combustible liquids
flash_point_f
159.8 °F - closed cup
flash_point_c
71 °C - closed cup
hcodes
Hazard Classifications
Skin Sens. 1A
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
