Skip to Content
Merck

1044298

USP

Atenolol Related Compound E

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,2′-{[(2-Hydroxypropane-1,3-diyl)bis(oxy)]bis(4,1-phenylene)}diacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H22N2O5
CAS Number:
Molecular Weight:
358.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

atenolol

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)Cc1ccc(cc1)OCC(O)COc2ccc(cc2)CC(=O)N

InChI

1S/C19H22N2O5/c20-18(23)9-13-1-5-16(6-2-13)25-11-15(22)12-26-17-7-3-14(4-8-17)10-19(21)24/h1-8,15,22H,9-12H2,(H2,20,23)(H2,21,24)

InChI key

RJTRBVLDVHIXNJ-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Atenolol Related Compound E USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Atenolol
  • Atenolol Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library