Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
glucosamine
manufacturer/tradename
USP
mp
211 °C (dec.) (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
InChI
1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8?/m1/s1
InChI key
OVRNDRQMDRJTHS-RTRLPJTCSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) USP Biologics |
| format neat | format neat | format neat | format neat |
| API family glucosamine | API family glucosamine | API family glucosamine | API family galactosamine |
| mp 211 °C (dec.) (lit.) | mp 190-194 °C (dec.) (lit.) | mp 211 °C (dec.) (lit.) | mp - |
Still not finding the right product?
Explore all of our products under N-Acetylglucosamine
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


