SU860076
Assembled screw cap with hole with PTFE/silicone septum
black polypropylene hole cap, thread for 8-425, PTFE/silicone (red PTFE/white silicone), septum thickness 1.3 mm, for use with 2 mL vial (standard opening), pkg of 100 ea
Synonym(s):
Screw Cap with PTFE/Silicone Septum
About This Item
Recommended Products
material
PTFE/silicone (red PTFE/white silicone)
black polypropylene hole cap
feature
closure type black polypropylene hole cap
hardness
45 (Durometer: shore A)
packaging
pkg of 100 ea
septum thickness
1.3 mm
fitting
thread for 8-425
compatibility
for use with 2 mL vial (standard opening)
Looking for similar products? Visit Product Comparison Guide
Application
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service