Skip to Content
Merck
All Photos(1)

Key Documents

RAD174

radiello Filtration Kit

pk of 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

packaging

pk of 20

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

General description

The filtration kit consists of graduated polyproylene syringe barrels and 13 mm diamater syringe filters with 0.45 μm porosity. The filtration kit is ideal for filtering aqueous samples prior to reversed-phase HPLC and ion-chromatography.

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Armelle Vallat et al.
Phytochemistry, 66(13), 1540-1550 (2005-06-14)
Headspace volatiles from apple-bearing twigs were collected in the field with a Radiello sampler during three different diurnal periods over the complete fruit growing season. Analyses by thermal desorption-GC-MS identified a total of 62 compounds in changing quantities, including the
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Rocco Rella et al.
The Science of the total environment, 487, 255-259 (2014-05-03)
The proofing treatment of the floor slabs with a solvent based material and successive treatment with epoxy resin resulted in significant air contamination in a primary school. The contamination caused illnesses in many occupants of school. The GC/MS investigations of
Anne Pennequin-Cardinal et al.
Talanta, 65(5), 1233-1240 (2008-10-31)
Radiello((R)) diffusive samplers filled with a thermally desorbable adsorbent (graphitised charcoal Carbograph 4) have been tested for the monitoring of BTEX. The sampling rates have been estimated under various controlled atmospheres in order to evaluate the effects of two factors

Articles

Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.

Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.

Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.

Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.

See All

Protocols

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service