PHL80016
(+)-Isomenthol
phyproof® Reference Substance
Synonym(s):
(1S,2R,5R)-(+)-Isomenthol, (1S,2R,5R)-2-Isopropyl-5-methylcyclohexanol
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (GC)
form
crystals
manufacturer/tradename
PhytoLab
bp
218-219 °C (lit.)
mp
77-83 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(C)[C@H]1CC[C@@H](C)C[C@@H]1O
InChI
1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9-,10+/m1/s1
InChI key
NOOLISFMXDJSKH-BBBLOLIVSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- (+)-Isomenthol for botanical discrimination: The study on Mentha plants using two-chiral column tandem GC-MS analysis of eight menthol enantiomers, including (+)-Isomenthol, underscores its utility in detailed botanical classification and the analysis of menthol enantiomers, aiding in the quality control and study of essential oils (Si et al., 2022).
- (+)-Isomenthol in food chemistry: Research focusing on the development of mass spectral libraries and gas chromatographic data for esters of diastereomeric menthols, including (+)-Isomenthol, offers valuable data for food chemistry applications, particularly in flavor analysis and food quality assessment (Boylan et al., 2021).
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service