Skip to Content
Merck
All Photos(1)

Key Documents

CRM64156

Supelco

Methoxychlor

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,1,1-Trichloro-2,2-bis-(p-methoxyphenyl)ethane, 2,2-Bis(4-methoxyphenyl)-1,1,1-trichloroethane, DMDT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15Cl3O2
CAS Number:
Molecular Weight:
345.65
Beilstein:
2057367
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

86-88 °C (lit.)

format

neat

SMILES string

COc1ccc(cc1)C(c2ccc(OC)cc2)C(Cl)(Cl)Cl

InChI

1S/C16H15Cl3O2/c1-20-13-7-3-11(4-8-13)15(16(17,18)19)12-5-9-14(21-2)10-6-12/h3-10,15H,1-2H3

InChI key

IAKOZHOLGAGEJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Methoxychlor is an organochlorine insecticide, used widely against a wide variety of insects in field crops, forage crops, vines, flowers, animal houses, dairies, and industries. It gets activated by contact and ingestion, modulating the voltage-gated sodium channels and causing hyperexcitation. It also exhibits endocrine-disrupting behavior by means of displaying both estrogenic and antiandrogenic activities.
According to the Commission Regulation (EC) No 2076/2002 of 20th November 2002, methoxychlor is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But a default maximum residue limit of 0.01-0.1 mg/kg is applicable for its presence in the products of plant and animal origin as per the EU Reg. 149/2008.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Methoxychlor CRM may also be used as given below:
  • To determine residues of selected pesticides in agricultural water samples by high-performance liquid chromatography (HPLC)(2)
    Ultrasound-assisted solvent extraction (UASE) and headspace solid-phase microextraction (HS-SPME) based extraction of 20 organochlorine pesticides from ethylenevinyl acetate (EVA) passive samplers employed for monitoring of marine surface waters, followed by a gas chromatography-tandem mass spectrometry (GC-MS/MS) detection
  • Simultaneous extraction and determination of 89 hydrophobic organic contaminants (HOCs) in marine sediment and biota samples using gas chromatography-electron ionization-triple quadrupole mass spectrometry (GC-EI-MS/MS) and liquid chromatography-electrospray ionization-triple quadrupole mass spectrometry (LC-ESI-MS/MS)
  • Determination of 26 organochlorine pesticides in tobacco samples using magnetic graphene for QuEChERS-based sample extraction and determination by online gel permeation chromatography-gas chromatography/tandem mass spectrometry (online GPC-GC-MS2)
  • Evaluate the coupling of gel-permeation clean-up (GPC) with gas-chromatography-tandem mass spectrometry (GC-MS/MS) and liquid-chromatography-tandem mass spectrometry (LC-MS/MS) techniques to determine various pesticides in fatty matrices
  • Multi-residue analysis of polycyclic aromatic hydrocarbons (PAHs) and organochlorine pesticides (OCPs) in personal product samples by gas chromatography in combination with flame ionization detector (GC-FID) and electron capture detector (GC-ECD)

Biochem/physiol Actions

Pesticide with estrogenic activity.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service