Skip to Content
Merck
All Photos(1)

Key Documents

CRM56578

Supelco

Prallethrin

mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(RS)-2-Methyl-4-oxo-3-prop-2-ynylcyclopent-2-enyl (1RS,3RS;1RS,3SR)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C/C1C(C(=O)OC2CC(=O)C(CC#C)=C2C)C1(C)C

InChI

1S/C19H24O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h1,9,14,16-17H,8,10H2,2-6H3

InChI key

SMKRKQBMYOFFMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Prallethrin is an insecticide that belongs to the family of non-fluorinated pyrethroids and is used against mosquitoes, houseflies, termites, and other insect vectors. It acts by modulating the sodium channels present across the axonal membranes in neurons.
As per the Regulation (EC) No. 1107/2009, repealing the directive 91/414/EEC, prallethrin is not approved for use as a plant protection agent in the European Union.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The prallethrin CRM may also find the following uses:

  • Determination of seven pyrethroids and piperonyl butoxide in indoor air following their passive sampling by a method combining thermal desorption with gas chromatography-mass spectrometry (GC-MS)
  • Multi-residue analysis of 8 pesticides in bovine, chicken, and swine muscle and liver samples by a method coupling modified QuEChERS extraction to LC-MS/MS
  • Analysis of 60 pesticide residues in various vegetable and fruit samples by solid-phase extraction (SPE) and ultra-high-performance liquid chromatography/time-of-flight mass spectrometry (UHPLC-TOF/MS)
  • Development of an electron ionization gas chromatography-mass spectrometry (GC-MS) method to determine l3 pyrethroid insecticide residues in whole blood samples
  • Simultaneous evaluation of nine pyrethroids in household pyrethroid products (mosquito coils and aerosols) by capillary gas chromatography with FID detection
  • Study the analysis of nine pyrethroids by combining hollow fiber-based liquid-phase microextraction (HF-LPME) with gas chromatography-mass spectrometry (GC/MS) in water samples

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I San Román et al.
Talanta, 100, 246-253 (2012-11-13)
A simple, easy-to-use, efficient and environmentally friendly method has been developed for the simultaneous analysis of nine pirethroid pesticides in water samples by the combination of hollow fibre-based liquid-phase microextraction (HF-LPME) and gas chromatography-mass spectrometry (GC/MS). For the developed method
K Hamamoto et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(7), 1162-1171 (2017-04-14)
Many multiresidue methods for the determination of pesticides in vegetables and fruits have been reported to date. However, few such methods have been employed to investigate pesticide residues in animal tissue. In this study, an LC-MS/MS multiresidue method coupled with
Insecticidal efficacy of a binary combination of cyphenothrin and prallethrin, applied as surface treatment against four major stored-product insects
Karanika C, et al.
Journal of Stored Products Research, 80, 41-49 (2019)
Atmakuru Ramesh et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 802(2), 371-376 (2004-03-17)
A new rapid and sensitive electron ionization gas chromatography-mass spectrometry method in selective ion monitoring mode (SIM) was developed for the determination of l3 synthetic pyrethroid insecticide molecules and their stereo isomers in whole blood. The pyrethroid insecticides investigated are
Caroline Raeppel et al.
Talanta, 131, 309-314 (2014-10-05)
A method coupling thermal desorption and gas chromatography/mass spectrometry (GC/MS) was developed for the simultaneous determination of 7 pyrethroids (allethrin, bifenthrin, cyphenothrin, imiprothrin, permethrin, prallethrin and tetramethrin) and piperonyl butoxide adsorbed on Tenax TA(®) passive samplers after exposure in indoor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service