Skip to Content
Merck
All Photos(1)

Key Documents

CRM40071

Supelco

Benzo[a]pyrene solution

certified reference material, TraceCERT®, 1000 μg/mL in acetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)cc3ccc4cccc5ccc2c3c45

InChI

1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H

InChI key

FMMWHPNWAFZXNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A P Wolterbeek et al.
Cancer letters, 89(1), 107-116 (1995-02-10)
Several experimental models have been developed to study respiratory tract carcinogenesis. The most widely applied in vivo model uses Syrian golden hamsters which receive intratracheal instillations of a suspension of benzo[a]pyrene (B[a]P) particles attached to ferric-oxide (Fe2O3) particles in saline;
Gunnar Boysen et al.
Mutation research, 543(1), 17-30 (2003-01-03)
We review studies which investigate the presence, using structure-specific analytical methods, of DNA or protein adducts of the carcinogen benzo[a]pyrene (BaP) in human tissues. The analytical methods include high performance liquid chromatography with fluorescence detection and gas chromatography-mass spectrometry. Although
F Mermillod-Blondin et al.
The Science of the total environment, 450-451, 230-241 (2013-03-19)
The interactions between invertebrates and micro-organisms living in streambed sediments often play key roles in the regulation of nutrient and organic matter fluxes in aquatic ecosystems. However, benthic sediments also constitute a privileged compartment for the accumulation of persistent organic
Susanna Sforzini et al.
Chemosphere, 107, 282-289 (2014-01-15)
An immunohistochemical method using antibodies against polycyclic aromatic hydrocarbons (PAHs) and dioxins was developed on frozen tissue sections of the earthworm Eisenia andrei exposed to environmentally relevant concentrations of benzo[a]pyrene (B[a]P) (0.1, 10, 50 ppm) and 2,3,7,8-tetrachloro-dibenzo-para-dioxin (TCDD) (0.01, 0.1
Kroum Alexandrov et al.
Toxicology letters, 198(1), 63-68 (2010-04-20)
Benzo(a)pyrene (BP) and cadmium are environmental pollutants found in foodstuffs, cigarette smoke, and polluted air. BP is converted in liver and lung to benzo(a)pyrene-7,8-diol-9,10-epoxide (BPDE) by the enzymes of the cytochrome P450 (CYP) superfamily, namely CYP1A1/1A2, and CYP1B1. BPDE reacts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service