Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
CRM18917
F.A.M.E. Mix
C14 - C22, certified reference material, wt. % (varied conc.), ampule of 100 mg
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
product line
TraceCERT®
form
liquid
CofA
current certificate can be downloaded
packaging
ampule of 100 mg
concentration
wt. % (varied conc.)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
multi-component solution
functional group
carboxylic acid
ester
shipped in
dry ice
storage temp.
-10 to -25°C
Application
Other Notes
Check out our complete portfolio of Fatty acid methyl ester (FAME) reference material solution mixes
Legal Information
Analyte
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.
GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.
GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.
GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
해당 제품에 들어있는 10가지 지방산의 구체적인 농도를 알고싶습니다
1 answer-
The specific concentration of each fatty acid will be reported on the lot specific Certificate of Analysis. Please see below for a link to a sample Certificate of Analysis: https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/rtc/CRM18917_LRAD0178.pdf
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service