BP1273
Mebeverine impurity C
British Pharmacopoeia (BP) Reference Standard
Synonym(s):
4-[Ethyl[2-(4-methoxyphenyl)-1-methylethyl]amino]-1-butanol
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
mebeverine
form
liquid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
N(C(Cc1ccc(cc1)OC)C)(CCCCO)CC
InChI
1S/C16H27NO2/c1-4-17(11-5-6-12-18)14(2)13-15-7-9-16(19-3)10-8-15/h7-10,14,18H,4-6,11-13H2,1-3H3
InChI key
ZGZAPRVKIAFOPL-UHFFFAOYSA-N
General description
Application
Also used in monographs such as:
Packaging
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service