Skip to Content
Merck
All Photos(1)

Key Documents

814472

TOYOPEARL® AF-Tresyl-650M Bulk Media

bottle of 100 g, phase tresyl-activated, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Tresyl-650M Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

dry, free flowing powder

packaging

bottle of 100 g

manufacturer/tradename

Tosoh 14472

extent of labeling

80 μmol/g dry resin ligand density

parameter

3 bar max. pressure

matrix

hydroxylated methacrylic polymer

matrix active group

tresyl-activated phase

particle size

65 μm

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Tresyl-650 resin is an activated resin which readily binds to amine and thiol groups. The resin is provided dry for introduction into a buffered solution containing protein or other ligand in neutral to slightly alkaline (pH 7-8) conditions.

Physical form

Shipped in dry, free flowing powder.

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service