Skip to Content
Merck
All Photos(1)

Key Documents

803410

Low Dead Volume (0.5 μm) HPLC PreColumn Filter

Stainless Steel Frit, pkg of 1 ea

Synonym(s):

TSKgel® Hardware and Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161807

Product Name

Low Dead Volume Precolumn Filter with 0.5 μm Stainless Steel Frit, pkg of 1 ea

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 03410

technique(s)

HPLC: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Low Dead Volume Precolumn Filter with 0.5 μm Stainless Steel Frit can be used to protect the TSKgel columns. When working with columns that contain smaller than 5 μm particles, Tosoh Bioscience suggests the installation of a pre-column filter in front of the (guard) column to avoid clogging the top frit. In addition, when suspecting debris from the breakdown of pump seals, it is beneficial to use a pre-injector membrane filter to prevent any particulates from reaching the injector and/or (guard) column.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Complete workflow for cannabis flower pesticide testing using LC-MS/MS and GC-MS/MS ensures product safety.

Complete workflow for cannabis flower pesticide testing using LC-MS/MS and GC-MS/MS ensures product safety.

Complete workflow for cannabis flower pesticide testing using LC-MS/MS and GC-MS/MS ensures product safety.

Complete workflow for cannabis flower pesticide testing using LC-MS/MS and GC-MS/MS ensures product safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service