Skip to Content
Merck
All Photos(1)

Key Documents

79491

Supelco

Metobromuron

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N′-(4-Bromophenyl)-N-methoxy-N-methylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O2
CAS Number:
Molecular Weight:
259.10
Beilstein:
2103964
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

SMILES string

CON(C)C(=O)Nc1ccc(Br)cc1

InChI

1S/C9H11BrN2O2/c1-12(14-2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)

InChI key

WLFDQEVORAMCIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Metobromuron is an herbicide active substance that belongs to the chemical group of phenylureas. It is a selective, systemic, pre-emergence herbicide used for the control of annual broad-leaved and grass weeds in beans, potatoes, tomatoes, tobacco, soybeans, maize, artichokes, and sugar beets.
It was approved on 1st January 2015 by the Commission Implementing Regulation (EU) No 890/2014 and is declared to be sanctioned under the Regulation (EC) No 1107/2009 as per Commission Implementing Regulation (EU) No 540/2011.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
The metobromuron CRM can also be used as given below:
  • Study the fate of sunflower herbicides, metobromuron and aclonifen in soil
  • Evaluate the photocatalytic degradation of metobromuron using TiO2 catalysts
  • Investigate metobromuron using selective electrochemical sensors in the presence of nanomaterials
  • Measure the levels of phenylurea herbicides in vegetables using SPME coupled with GC
  • Determine selected pesticides from tobacco using high-performance liquid chromatography-electrospray ionisation-tandem mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karolina Sipa et al.
Analytica chimica acta, 1030, 61-69 (2018-07-24)
In the present work, bare ultra trace graphite electrode (UTGE), UTGE modified with multi-walled carbon nanotubes (UTGE-MWCNTs), and UTGE modified with graphene nanoplatelets (UTGE-GNPs) were considered as working electrodes. For the first time, the UTGEs were modified with MWCNTs and
H Berrada et al.
Journal of chromatography. A, 1042(1-2), 9-14 (2004-08-07)
Residues of metobromuron, monolinuron and linuron herbicides and their aniline homologous were analyzed in carrots, onions and potatoes by solid-phase microextraction (SPME) performed with a polyacrylate fiber. A juice was obtained from food samples that were further diluted, and an
Bernhard Mayer-Helm et al.
Talanta, 74(5), 1184-1190 (2008-03-29)
A method was developed for the quantitative determination of alachlor, benalaxyl, clomazone, diflubenzuron, dimethomorph, diphenamid, ethofumesate, metalaxyl, methoprene, metobromuron and piperonyl butoxide on tobacco. The pesticides were extracted with water and methanol from five different types of tobacco. The extracts
Phototransformation of metobromuron in the presence of TiO2
Amine-Khodja A, et al.
Applied Catalysis. B, Environmental, 147-154 null
Behaviour of two sunflower herbicides (metobromuron, aclonifen) in soil
Vischetti C, et al.
Chemosphere, 231-238 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service