59680-U
Supel™ BioSPME 96-Pin Device
pack of 1 ea
Synonym(s):
Supel™ BioSPME 96-Pin Device
About This Item
Recommended Products
packaging
pack of 1 ea
Quality Level
technique(s)
solid phase microextraction (SPME): suitable
matrix active group
C18 coating
application(s)
clinical
pharmaceutical
compatibility
for use with automated robotic liquid handling systems
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Determination of plasma protein binding using a SupelTM C18 pin device based BioSPME method for efficient sample preparation, along with a comparison to the equilibrium dialysis method.
Determination of plasma protein binding using a SupelTM C18 pin device based BioSPME method for efficient sample preparation, along with a comparison to the equilibrium dialysis method.
Determination of plasma protein binding using a SupelTM C18 pin device based BioSPME method for efficient sample preparation, along with a comparison to the equilibrium dialysis method.
Determination of plasma protein binding using a SupelTM C18 pin device based BioSPME method for efficient sample preparation, along with a comparison to the equilibrium dialysis method.
Related Content
Supel™ BioSPME 96-Pin devices enhance speed and simplicity for plasma protein binding studies and LCMS analyses.
Supel™ BioSPME 96-Pin devices enhance speed and simplicity for plasma protein binding studies and LCMS analyses.
Supel™ BioSPME 96-Pin devices enhance speed and simplicity for plasma protein binding studies and LCMS analyses.
Supel™ BioSPME 96-Pin devices enhance speed and simplicity for plasma protein binding studies and LCMS analyses.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service