Skip to Content
Merck
All Photos(3)

Key Documents

57344-U

Supelco

SPME Fiber Assembly

greener alternative

CAR/PDMS, for use with manual holder, Nitinol-core (NIT), 23 ga, 75 μm

Synonym(s):

Carboxen/PDMS SPME fiber assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21
Pricing and availability is not currently available.

Product Name

SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder

material

fused silica fiber
plain black hub

needle size

23 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

75 μm (CAR/PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

CAR/PDMS coating

application(s)

food and beverages

compatibility

(For gasses and low molecular weight compounds (MW 30 - 225).)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

For Gases and Low Molecular Weight Compounds (MW 30-225)
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application


  • Evaluation of a multi-fiber exchange solid-phase microextraction system and its application to on-site sampling: This study introduces a multi-fiber exchange system for solid-phase microextraction, specifically showcasing the efficiency of the Carboxen/Polydimethylsiloxane (CAR/PDMS) fibers in on-site sampling scenarios. The application emphasizes rapid and adaptable sampling methods that cater to dynamic environmental and laboratory conditions, enhancing both the practicality and scope of SPME′s utility in the field (Gómez-Ríos GA, Reyes-Garcés N, Pawliszyn J, 2015).

Footnote

This SPME fiber assembly is also available with a Nitinol (NIT) core as Cat. No. 57901-U, providing enhanced fiber reproducibility, greater fiber durability, maintained selectivity for method consistency, and core inertness to ensure analyte stability. See more at SigmaAldrich.com/NITSPME .

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T R Dubie et al.
Journal of medical entomology, 54(5), 1299-1304 (2017-04-12)
Commercial livestock facilities are faced with the challenge of managing large amounts of waste including manure and animal mortalities. One method of disposing of dead animals is composting. The cadavers are enveloped in carbon material that creates a barrier between
Jérémy Ratel et al.
Journal of chromatography. A, 1216(45), 7889-7898 (2009-09-29)
Animal-derived products are particularly vulnerable to contamination by volatile organic compounds (VOCs). These lipophilic substances, which are generated by an increasing number of sources, are easily transferred to the atmosphere, water, soil, and plants. They are ingested by livestock and
E Serrano et al.
Journal of chromatography. A, 1216(1), 127-133 (2008-11-29)
The objective of this paper is to investigate the potential of multiple headspace-solid-phase microextraction (MHS-SPME) for the determination of volatile compounds in complex matrix samples. A method based on MHS-SPME for the determination of around 20 volatile compounds, responsible of
Sudhir Kumar Pandey et al.
Journal of chromatography. A, 1216(28), 5436-5444 (2009-06-06)
In this study, the feasibility of GC-MS was evaluated for the quantification of odorous volatile organic compounds (VOCs) in environmental samples. These included methyl ethyl ketone, isobutyl alcohol, methyl isobutyl ketone, and butyl acetate plus benzene, toluene, and xylene (BTX).
Carlo Bicchi et al.
Journal of chromatography. A, 1218(6), 753-762 (2011-01-05)
The study compares standard addition (SA), stable isotope dilution assay (SIDA) and multiple headspace extraction (MHE) as methods to quantify furan and 2-methyl-furan in roasted coffee with HS-SPME-GC-MS, using CAR-PDMS as fibre coating, d(4)-furan as internal standard and in-fibre internal

Questions

1–4 of 4 Questions  
  1. Would this Carboxen/Polydimethylsiloxane (CAR/PDMS) fibre be suitable for the extraction of carbon monoxide?

    1 answer
    1. Carbon Monoxide is very likely too small to be retained. There has been some feedback that CO2 was detected in GC/MS using Carbowax/Divinylbenzene SPME fibers, however this has not been validated. A CW/DVB fiber option is not offered. Please check the literature for further information.

      Helpful?

  2. Is SPME fiber assembly Carboxen/Polydimethylsiloxane(CAR/PDMS), Product 57344-U, also available with the new metal core technology?

    1 answer
    1. Yes, this fiber is available with a fused silica, Stableflex, or metal fiber cores.  The metal fiber assemblies are only available in 23 gauge needles, and should be used with a Merlin Microseal on the GC injection port

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What types of compounds would be suitable for extraction using SPME fiber assembly Carboxen/Polydimethylsiloxane(CAR/PDMS), Product 57344-U?

    1 answer
    1. This fiber is best suited for extracting relatively small volatile compounds.  The approximate size range of the compounds is between 2-10 carbons, and should be suitable for most functional groups (i.e., alcohols, acids, mercaptans, etc).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service