Skip to Content
Merck
All Photos(2)

Documents

57333-U

Supelco

SPME Sampling Stand

greener alternative

for use with 4 mL vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

solid phase microextraction (SPME): suitable

application(s)

food and beverages

compatibility

for use with 4 mL vials

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
Holds eight vials while supporting the SPME syringe for consistent fiber immersion depth. Cat. No. 57333-U accommodates 4 mL vials only; Cat. No. 57357-U accommodates 15 mL vials. Order the 15 mL vial puck (Cat. No. 57358-U) as a replacement for the 15 mL unit, or to use 15 mL vials with the 4 mL unit. Not for use with automated/HPLC fiber holders.

recommended

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Tytgat et al.
International journal of legal medicine, 109(3), 150-154 (1996-01-01)
The most common alkyl nitrites encountered in forensic toxicology are iso-butyl, n-butyl and iso-pentyl(amyl) nitrites. All have become popular as an aphrodisiac, especially among the homosexual population. Alkyl nitrites are a volatile and unstable group of compounds, which hydrolyse in
J H Han et al.
Journal of food science, 73(8), E349-E355 (2008-11-21)
Physical properties of whey protein isolate (WPI) coating solution incorporating ascorbic palmitate (AP) and alpha-tocopherol (tocopherol) were characterized, and the antioxidant activity of dried WPI coatings against lipid oxidation in roasted peanuts were investigated. The AP and tocopherol were mixed
Alfredo Koch et al.
Phytochemistry, 71(17-18), 2190-2198 (2010-10-23)
2-Methoxy-3-isobutylpyrazine (MIBP) contributes a bell pepper aroma to many grape cultivars and has a reported aroma threshold of ∼2 ng L(-1) in water. The purpose of this study was twofold: (1) develop a procedure using headspace solid phase micro-extraction combined
A P Arisseto et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(12), 1832-1839 (2012-08-23)
Selected commercial processed foods available in the Brazilian market (306 samples) were analysed for furan content using a validated gas chromatography-mass spectrometry method preceded by headspace solid phase micro-extraction (HS-SPME-GC/MS). Canned and jarred foods, including vegetable, meat, fruit and sweet
Marghereth Saverini et al.
Journal of photochemistry and photobiology. B, Biology, 108, 8-15 (2012-01-20)
The presence of (±)α-pinene, (+)β-pinene, (+)3-carene, and R-(+)limonene terpenes in wastewater of a citrus transformation factory was detected and analyzed, in a previous study, by using Solid Phase Micro-extraction (SPME) followed by GC analyses. Purpose of that research was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service