57037
Visiprep™ Manifold Collection Rack (included with standard and DL models)
for use with 12-port model, includes base, 3 support rods, center plate, plate for 10 mm test tubes, 12 retaining clips
Synonym(s):
V.M. COLLECTION RACK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 12-port model
Looking for similar products? Visit Product Comparison Guide
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Michael S Gardner et al.
Analytical chemistry, 81(2), 557-566 (2008-12-17)
Defensins are highly basic cationic peptides that are important components of the innate and adaptive immune response pathways. In addition, these peptides are involved in CD8+ T cell response to HIV-1, increased pulmonary infection risk among cystic fibrosis patients, upregulated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service