56107
Frit for Omnifit® Column Endpiece
PTFE, for use with 10 mm bore, pore size 10 μm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE
packaging
pkg of 20 ea
manufacturer/tradename
Omnifit 6146
technique(s)
LPLC: suitable
pore size
10 μm
compatibility
for use with 10 mm bore
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
End pieces are equipped with porous frits. These can be made up of sintered metal, glass and plastic. The pore diameter and surface structures of the frits should be optimised such as to avoid clogging and abrasion of the chromatographic particles. The frits are made such that, they are easily exchangeable. Frits can also be used to keep the packing material in place.
Application
Polyethylene frits may be used for glass columns to conduct filtration experiments with bacterial suspensions. PTFE frits may be used for preparative chromatography and solid phase extraction for borosilicate columns.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Subperiosteal osteoid osteoma of the distal phalanx of the fourth toe.
Orthopedics, 24(7), 695-696 (2001-08-02)
Journal of chromatography. A, 887(1-2), 43-53 (2000-08-29)
Considering the current interest in capillary electrochromatography (CEC), performed in packed columns, we present the different methods used to pack capillary columns for use in CEC. General considerations on column packing are given and the column fabrication process is discussed
Protein Purification: Principles, High Resolution Methods, and Applications, 19-19 (2012)
Population heterogeneity affects transport of bacteria through sand columns at low flow rates.
Environmental Science & Technology, 32, 2100-2105 (1998)
Improvements to the pretreatment of bone at Oxford.
Radiocarbon, 46, 155-164 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service